Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for 96 Deep-Well Plates or 1 3/4" Max H Boxes, 6 x 7 Array
Catalog No. 501960741 Shop All Cole-Parmer Products
Encompass_Preferred
With these freezer drawer racks, you can easily access fewer microtiter plates and boxes at a time, while at the same time protecting other samples in the rack from warming up. They are made of durable corrosion-resistant stainless steel for long-term use
Catalog No. 50-196-0741 Supplier Cole-Parmer Supplier No. 0440081
Add to Cart
Add to Cart
With these freezer drawer racks, you can easily access fewer microtiter plates and boxes at a time, while at the same time protecting other samples in the rack from warming up. They are made of durable corrosion-resistant stainless steel for long-term use