Learn More
Cole-Parmer Upright Freezer Bin Rack Large, with Lid 26.54 x 8.86 x 5.43" (L x H x W)
Catalog No. NC1434658 Shop All Cole-Parmer Products
Encompass_Preferred
Organize and Optimize your Laboratory FreezerPolarSafe Upright Freezer Bin Racks are are designed to store miscellaneous bottles, tubes, vials, bags or boxes. Racks sides are made from wire mesh to allow for easy viewing of contents, the open format works well with -20C freezers. Racks are available in both an upright (including lid) and drawer style. The drawer style racks are fully framed and work like standard drawer racks. All Upright Freezer Bin racks are constructed of high quality corrosion resistant stainless steel.
Catalog No. NC1434658 Supplier Cole-Parmer Supplier No. 0439784
Organize and Optimize your Laboratory FreezerPolarSafe Upright Freezer Bin Racks are are designed to store miscellaneous bottles, tubes, vials, bags or boxes. Racks sides are made from wire mesh to allow for easy viewing of contents, the open format works well with -20C freezers. Racks are available in both an upright (including lid) and drawer style. The drawer style racks are fully framed and work like standard drawer racks. All Upright Freezer Bin racks are constructed of high quality corrosion resistant stainless steel.