Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack, 2 Drawers for 6 Boxes & 1 for (30) 50 mL Tubes
Catalog No. 501960758 Shop All Cole-Parmer Products
Encompass_Preferred
These specialized drawer racks lets you store 15 or 50-mL centrifuge tubes and storage bottles without a box. Drawer racks provide access to samples without having to remove the entire rack from the freezer.
Catalog No. 50-196-0758 Supplier Cole-Parmer Supplier No. 0440156
These specialized drawer racks lets you store 15 or 50-mL centrifuge tubes and storage bottles without a box. Drawer racks provide access to samples without having to remove the entire rack from the freezer.