Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack, 2 Drawers for 6 Boxes & 1 Drawer for Bottles
Catalog No. 501960756 Shop All Cole-Parmer Products
Encompass_Preferred
These specialized drawer racks lets you store 15 or 50-mL centrifuge tubes and storage bottles without a box. Drawer racks provide access to samples without having to remove the entire rack from the freezer.
Catalog No. 50-196-0756 Supplier Cole-Parmer Supplier No. 0440146
These specialized drawer racks lets you store 15 or 50-mL centrifuge tubes and storage bottles without a box. Drawer racks provide access to samples without having to remove the entire rack from the freezer.