Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for 96/384-Well Plates, 96-Well Boxes, or Boxes w/ 2 1/8" Max H, 5 x 3 Array
Catalog No. 501960730 Shop All Cole-Parmer Products
Encompass_Preferred
Quickly store and retrieve plates and boxes without taking out the whole rack using these freezer drawer racks. They accommodate microtube boxes with 2-1/8-inch maximum height, and made of durable corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0730 Supplier Cole-Parmer Supplier No. 0440028
Quickly store and retrieve plates and boxes without taking out the whole rack using these freezer drawer racks. They accommodate microtube boxes with 2-1/8-inch maximum height, and made of durable corrosion-resistant stainless steel for long-term use.