Learn More
Cole-Parmer FRZR DRWR RCK 2 BOX 3X3 9BOXES
Catalog No. NC0525504 Shop All Cole-Parmer Products
Encompass_Preferred
Upright Freezer Drawer Rack for 2 in. Boxes, 3 x 3 capacity 9 boxes 16-1/2 x 7-1/8 x 5-1/2
Catalog No. NC0525504 Supplier Cole-Parmer Supplier No. 0439819
Upright Freezer Drawer Rack for 2 in. Boxes, 3 x 3 capacity 9 boxes 16-1/2 x 7-1/8 x 5-1/2