Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for Standard 2" Boxes, 4 x 3 Array
Catalog No. 501960497 Shop All Cole-Parmer Products
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer. Choose from top or side loading depending on application requirements.
Catalog No. 50-196-0497 Supplier Cole-Parmer Supplier No. 0439853
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer. Choose from top or side loading depending on application requirements.