Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Eco-Rack for Standard 2" Boxes, 3 x 4 Array
Catalog No. 501960533 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer, Eco-racks is an economical alternative to standard racks and rreat for storing standard 2" cardboard, plastic,or metal boxes Designed to fit major brands of upright freezers, they are crafted of sturdy corrosion-resistant stainless steel.
Catalog No. 50-196-0533 Supplier Cole-Parmer Supplier No. 0440351
These upright freezer, Eco-racks is an economical alternative to standard racks and rreat for storing standard 2" cardboard, plastic,or metal boxes Designed to fit major brands of upright freezers, they are crafted of sturdy corrosion-resistant stainless steel.