Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Eco-Rack for Standard 2" Boxes, 4 x 3 Array
Catalog No. 501960792 Shop All Cole-Parmer Products
Encompass_Preferred
These eco-racks are great for storing standard two-inch cardboard, plastic or metal boxes. Designed to fit major brands of upright freezers, the racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0792 Supplier Cole-Parmer Supplier No. 0440354
These eco-racks are great for storing standard two-inch cardboard, plastic or metal boxes. Designed to fit major brands of upright freezers, the racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.