Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for 100-Cell Hinged Plastic Boxes, 12 box capacity
Catalog No. 501960721 Shop All Cole-Parmer Products
Encompass_Preferred
These racks come with locking rods to securely store multiple 100-cell hinged top plastic boxes in place. The racks have sturdy dual handles for easy handling, and are made of corrosion-resistant stainless steel.
Catalog No. 50-196-0721 Supplier Cole-Parmer Supplier No. 0439981
These racks come with locking rods to securely store multiple 100-cell hinged top plastic boxes in place. The racks have sturdy dual handles for easy handling, and are made of corrosion-resistant stainless steel.