Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for Standard 2" Boxes, 15 box capacity
Catalog No. 501960695 Shop All Cole-Parmer Products
Encompass_Preferred
These racks come with locking rods to securely keep boxes in place. They accommodate standard two-inch cardboard, plastic and metal boxes. The racks have dual stainless steel handles for easy handling, and are made of sturdy corrosion-resistant stainless steel.
Catalog No. 50-196-0695 Supplier Cole-Parmer Supplier No. 0439881
These racks come with locking rods to securely keep boxes in place. They accommodate standard two-inch cardboard, plastic and metal boxes. The racks have dual stainless steel handles for easy handling, and are made of sturdy corrosion-resistant stainless steel.