Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for Mini 2" Boxes, 11 box capacity
Catalog No. 501960699 Shop All Cole-Parmer Products
Encompass_Preferred
These freezer racks are great for storing mini two-inch cardboard or plastic boxes. A locking rod securely holds boxes in place. The racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0699 Supplier Cole-Parmer Supplier No. 0439896
These freezer racks are great for storing mini two-inch cardboard or plastic boxes. A locking rod securely holds boxes in place. The racks are crafted of sturdy corrosion-resistant stainless steel for long-term use.