Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for Standard 3" Boxes, 8 box capacity
Supplier:  Cole-Parmer 0439947
Encompass_Preferred
These racks come with locking rods to securely keep boxes in place. They accommodate standard three-inch cardboard, plastic and metal boxes. The racks have dual metal handles for easy handling, and are made of sturdy corrosion-resistant stainless steel.
Catalog No. 50-196-0711