Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical Freezer Rack for 96 Deep-Well Microtiter Plates or Boxes w/ 1 3/4" Max H, 12 plate or box capacity
Catalog No. 501960747 Shop All Cole-Parmer Products
Encompass_Preferred
These racks come with locking rods to securely store microtiter plates or boxes in place. The racks have sturdy dual handles for easy handling, and made of corrosion-resistant stainless steel.
Catalog No. 50-196-0747 Supplier Cole-Parmer Supplier No. 0440116
Add to Cart
Add to Cart
These racks come with locking rods to securely store microtiter plates or boxes in place. The racks have sturdy dual handles for easy handling, and made of corrosion-resistant stainless steel.