Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Large 2" Boxes & 100-Cell Cryoboxes, 4 x 6 Array
Catalog No. 501960717 Shop All Cole-Parmer Products
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
Catalog No. 50-196-0717 Supplier Cole-Parmer Supplier No. 0439976
Add to Cart
Add to Cart
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.