Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Standard 2" Boxes, 2 x 3 Array
Catalog No. 501960692 Shop All Cole-Parmer Products
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer.
Catalog No. 50-196-0692 Supplier Cole-Parmer Supplier No. 0439873
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer.