Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96/384-Well Microtiter Plates, accepts 96 with lids or 112 without lids
Catalog No. 501960723 Shop All Cole-Parmer Products
Encompass_Preferred
These racks come with locking rods to securely store microtiter plates in place. The racks have sturdy dual handles for easy handling, and are made of corrosion-resistant stainless steel.
Catalog No. 50-196-0723 Supplier Cole-Parmer Supplier No. 0439991
These racks come with locking rods to securely store microtiter plates in place. The racks have sturdy dual handles for easy handling, and are made of corrosion-resistant stainless steel.