Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96/384-Well Microtiter Plates, accepts 96 with lids or 120 without lids
Catalog No. 501960725 Shop All Cole-Parmer Products
Encompass_Preferred
These racks come with locking rods to securely store microtiter plates in place. The racks have sturdy dual handles for easy handling, and are made of corrosion-resistant stainless steel.
Catalog No. 50-196-0725 Supplier Cole-Parmer Supplier No. 0439999
These racks come with locking rods to securely store microtiter plates in place. The racks have sturdy dual handles for easy handling, and are made of corrosion-resistant stainless steel.