Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96/384-Well Microtiter Plates, accepts 112 with lids or 140 without lids
Catalog No. 501960509 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks conveniently store microtiter plates. They have locking rods to keep plates in place. The racks are flanked with handles for quick storage and retrival of rack. They are crafted of corrosion-resistent stainless steel for durablity.
Catalog No. 50-196-0509 Supplier Cole-Parmer Supplier No. 0440009
These upright freezer racks conveniently store microtiter plates. They have locking rods to keep plates in place. The racks are flanked with handles for quick storage and retrival of rack. They are crafted of corrosion-resistent stainless steel for durablity.