Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96 Deep-Well Microtiter Plates or 1 3/4" Max H Boxes, 5 x 5 Array
Catalog No. 501960511 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks conveniently store microtiter plates and microtube boxes. They are crafted of corrosion-resistent stainless steel for durablity.
Catalog No. 50-196-0511 Supplier Cole-Parmer Supplier No. 0440047
These upright freezer racks conveniently store microtiter plates and microtube boxes. They are crafted of corrosion-resistent stainless steel for durablity.