Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96 Deep-Well Microtiter Plates or 1 3/4" Max H Boxes, 6 x 4 Array
Catalog No. 501960736 Shop All Cole-Parmer Products
Encompass_Preferred
Safely store large quantities of 96 deep-well microtiter plates and microtube boxes with these freezer racks. They come with a locking rod to securely keep them in place. And, you can access them from either side of the rack -- accomodating freezers with left- or right-handed doors.
Catalog No. 50-196-0736 Supplier Cole-Parmer Supplier No. 0440049
Safely store large quantities of 96 deep-well microtiter plates and microtube boxes with these freezer racks. They come with a locking rod to securely keep them in place. And, you can access them from either side of the rack -- accomodating freezers with left- or right-handed doors.