Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96-Well Microtube Boxes or Boxes w/ 2 1/8" Max H, 6 x 4 Array
Catalog No. 501960743 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks come with a locking rod to securely hold boxes in place. Boxes can be accessed from either side of rack, which has handles on both sides to accommodate freezers with left- or right-handed doors. They are crafted of corrosion-resist stainless steel for long-term use.
Catalog No. 50-196-0743 Supplier Cole-Parmer Supplier No. 0440100
These upright freezer racks come with a locking rod to securely hold boxes in place. Boxes can be accessed from either side of rack, which has handles on both sides to accommodate freezers with left- or right-handed doors. They are crafted of corrosion-resist stainless steel for long-term use.