Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 96-Well Microtube Boxes or Boxes w/ 2 1/8" Max H, 7 x 4 Array
Catalog No. 501961832 Shop All Cole-Parmer Products
Encompass_Preferred
Freezer includes locking rods that securely hold boxes in place. Access boxes from either side of rack Handles on both sides accommodate freezers with left- or right-handed door Sturdy corrosion-resistant stainless steel construction for long-term use
Catalog No. 50-196-1832 Supplier Cole-Parmer Supplier No. 0440105
Freezer includes locking rods that securely hold boxes in place. Access boxes from either side of rack Handles on both sides accommodate freezers with left- or right-handed door Sturdy corrosion-resistant stainless steel construction for long-term use