Learn More
Foxx Life Sciences EZBiopure Feed Systems, 500mL Bottle Top Filter, 0.22µm Membrane, PP Natural GL45 VersaCap with adapter 2x 1/8 HB with diptube, EZBio TPE Tubing, AQS HT and Vented, 4/CS
Supplier:  Foxx Life Sciences 5C69230FLS
Encompass_Preferred
Foxx Life Sciences EZBiopure Feed System, a Single-Use solution designed for Filtration, Fluid Transfer, and Storage in Bioprocessing. This system combines the Autofil 2 Filtration Unit, EZBiopure TR Media Bottles, and TR Cap Assembly. All components are molded from USP Class VI materials and undergo Gamma Irradiation
Autofil 2 Bottle Top Filtration Unit features a 0.2μm PES membrane, ideal for media prep and transfer to Bioreactors. It filters substances like FBS and glycerol, available in 250mL, 500mL,1L .
EZBiopure TR Media Bottles are made of Polycarbonate, GL45 neck and molded finger grips for easy handling. Raised graduation marks ensure accurate dispensing.
EZBiopure TR Cap Assembly has two ports vent filter and Connector Fitting with Pinch Clamp for flow control.
Applications
Media and buffer transfer
Bioprocess Product sampling and supplement injection
Product storage
Filtration and purification process connection
Cell harvesting
Catalog No. 50-255-8689