Learn More
MSC ZEP Foaming Acid Cleaner, Liquid, 1 gal Bottle, 4/case
Catalog No. 502815787
Encompass
ZEP FS.CONC Foaming Acid Cleaner is a powerful liquid solution designed for heavy-duty cleaning in food and beverage facilities. It effectively removes stubborn deposits and grime.
  • Formulated with orthophosphoric and sulphuric acids for robust cleaning
  • Exhibits a low pH (<1), indicating strong acidic properties
  • Clear amber liquid with a characteristic odor
  • Soluble in water for easy application and rinsing
  • Recommended for food and beverage facility maintenance
  • Supplied in 1 gal. bottles, 4 ea/case
Catalog No. 50-281-5787 Supplier MSC Supplier No. 17641069
ZEP FS.CONC Foaming Acid Cleaner is a powerful liquid solution designed for heavy-duty cleaning in food and beverage facilities. It effectively removes stubborn deposits and grime.
  • Formulated with orthophosphoric and sulphuric acids for robust cleaning
  • Exhibits a low pH (<1), indicating strong acidic properties
  • Clear amber liquid with a characteristic odor
  • Soluble in water for easy application and rinsing
  • Recommended for food and beverage facility maintenance
  • Supplied in 1 gal. bottles, 4 ea/case