Learn More
Roche Diagnostics FS ESSENTIAL DNA GREEN MASTER
Supplier:  Roche Diagnostics 06402712001
Encompass
FastStart Essential DNA Green Master Ready-to-use hot start reaction mix for SYBR Green I-based real-time PCR using the LightCycler 96 Instrument and the LightCycler Nano Instrument. 5 x 1 ml for up to 500 reactions of 20 l final volume
Catalog No. 50-588-375