Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC FS FORMULA 12167 CLEANER

Catalog No. 502818459
Encompass

Zep FS Formula 12167 is a chlorinated disinfectant, sanitizer, and deodorant specifically formulated for use in food processing plants and retail establishments. It contains 5.25% available chlorine, effectively sanitizing various surfaces and equipment.

  • Contains 5.25% available chlorine
  • USDA applicable (D2) and EPA registered
  • Suitable for egg shell and commercial laundry sanitization
  • Effective on hard nonporous surfaces
  • Requires no rinsing after use
  • Liquid form with a strong chlorine odor
  • Ph range of 12.5–13.0
  • Density of 1.02 g/cm3

Catalog No. 50-281-8459 Supplier MSC Supplier No. 17641572
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zep FS Formula 12167 is a chlorinated disinfectant, sanitizer, and deodorant specifically formulated for use in food processing plants and retail establishments. It contains 5.25% available chlorine, effectively sanitizing various surfaces and equipment.

  • Contains 5.25% available chlorine
  • USDA applicable (D2) and EPA registered
  • Suitable for egg shell and commercial laundry sanitization
  • Effective on hard nonporous surfaces
  • Requires no rinsing after use
  • Liquid form with a strong chlorine odor
  • Ph range of 12.5–13.0
  • Density of 1.02 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.