Learn More
Cic FS100 ALUMINUM SEALS
Catalog No. NC9327419
Encompass_Preferred
Seal; Foil; Aluminum; Certified Rnase/Dnase Free; Suitable for -80deg.C Storage
Catalog No. NC9327419 Supplier Cic Supplier No. FS100
Seal; Foil; Aluminum; Certified Rnase/Dnase Free; Suitable for -80deg.C Storage