Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FSB, ≥95%, Calbiochem™,

Catalog No. 3441015MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-410-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-410-15MG Supplier MilliporeSigma™ Supplier No. 3441015MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 760988-03-2
Molecular Formula C24H17FO6
Synonym (E,E)-1-Fluoro-2,5-bis(3-hydroxycarbonyl-4-hydroxy)styrylbenzene

Specifications

Packaging Glass Bottle
Color Yellow
Quantity 5 mg
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.