Learn More
Leica Biosystems FSC 22 BLUE 4OZ 9/CS
Catalog No. NC0278476
Encompass_Preferred
FSC 22 is a water soluble embedding compound used in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. It provides excellent sectioning consistency with minimal curling of sections at a working temperature of -20 °C. FSC 22 is available in clear or in light blue for better visualization of small specimens. It is highly recommended for surgical pathology laboratories.
Catalog No. NC0278476 Supplier Leica Biosystems Supplier No. 3801481
FSC 22 is a water soluble embedding compound used in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. It provides excellent sectioning consistency with minimal curling of sections at a working temperature of -20 °C. FSC 22 is available in clear or in light blue for better visualization of small specimens. It is highly recommended for surgical pathology laboratories.