Learn More
Fisherbrand™ FSC Chemical Absorbent Pads
Catalog No. 19140665 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
17 gal.
Catalog No. Capacity (English)
19-140-665 17 gal.
1 options
Catalog No. 19-140-665
  • $187.00
    $154.65 / Case of 100
    Save $32.35
    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste. Ideal in areas with a high risk of slips, trips and falls.
  • High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste. Ideal in areas with a high risk of slips, trips and falls.
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert.
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material.
  • High Visibility Series feature safety signage and absorbency in one.
  • Perforations provide cost control and reduce waste by allowing you to use only what you need. Printed Message: Warns of slips, trips and falls.
  • Bright Color: Warns of presence of leaks, drips or spills even in low light.
Specifications
Capacity (English) Sorbing 17 gal.
Color Yellow
Capacity (English) 17 gal.
Material Polypropylene
Type Spill Control Pads
Includes 100 Pads/Cs.
Dimensions (L x W) 19 x 15 in.