Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ FSC Drum Spill Control Kit - Universal Application

Catalog No. 19140664 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English) Sorbing:
5 gal.
17 gal.
41 gal.
75 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Sorbing
19-140-664 17 gal.
19-140-662 5 gal.
19-140-658 75 gal.
19-140-661 41 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-140-664
  • $325.00
    $251.65 / Each of 1

    Save $73.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spill Kits combine absorbents, accessories, PPE and instructional guidelines so you are prepared for emergencies when a spill occurs.

  • Spill Kits combine absorbents, accessories, PPE and instructional guidelines so you are prepared for emergencies when a spill occurs.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids.
  • UN-approved, tough, secure, screw-topped drum that is ideal for large spills.
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills. Spill kits are ideal for fast response to critical accidents.

Specifications

Capacity (English) Sorbing 17 gal.
Color Gray
Material Polypropylene
Type Spill Control Kit
Includes (12) 15 x 19 in. Pads;(2) 18 x 18 in. Pillows;(3) 3 in. x 12 ft. Socks;(3) Disposal Bags; Goggles; Handbook; Nitrile Gloves
Dimensions (L x W) 21 x 18 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.