Learn More
Fisherbrand™ FSC High Visibility Safety Absorbent Pads
Catalog No. 19140668 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English) Sorbing:
22 gal.
Catalog No. Capacity (English) Sorbing
19-140-668 22 gal.
1 options
Catalog No. 19-140-668
High-Visibility Absorbents combine effective absorbency and hazardous-materials messaging in one convenient solution.
  • High-Visibility Absorbents combine effective absorbency and hazardous-materials messaging in one convenient solution.
  • Surfactant-treated polypropylene picks up a wide range of aggressive chemicals, including hydrofluoric acid.
  • Bright color and printed message alert workers to the presence of hazardous materials and waste.
  • Green coverstock identifies product for proper separation and disposal, and reduces likelihood of linting or tearing.
  • Heavy weight: specifically designed for continuous leaks and larger spills so you use less material.
  • Perforations reduce waste by allowing you to use only what you need.
Specifications
Capacity (English) Sorbing 22 gal.
Color Green
Capacity (English) 22 gal.
Material Polypropylene
Type Spill Control Pads
Includes 100 Pads/Cs.
Dimensions (L x W) 19 x 15 in.