Learn More
Fisherbrand™ FSC Special Spill Kit Formaldehyde
Catalog No. 19140666 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English) Sorbing:
4.5 gal.
Catalog No. Capacity (English) Sorbing
19-140-666 4.5 gal.
1 options
Catalog No. 19-140-666
  • $281.00
    $247.65 / Each of 1
    Save $33.35
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Spill Kits combine pads, accessories and PPE so that you are prepared in an emergency. Position them around your facility so that you are prepared should a spill occur.
  • Spill Kits combine pads, accessories and PPE so that you are prepared in an emergency.
  • Position them around your facility so that you are prepared should a spill occur.
  • Spill kit contains surfactant-treated absorbents and contents designed to pick up formaldehyde while remaining chemically inert.
  • Reusable screw-top bucket keeps contents clean and easily accessible. Absorbs up to 4.5 gallons per kit.
Specifications
Capacity (English) Sorbing 4.5 gal.
Color Green
Material Polypropylene
Type Spill Control Kit
Includes (1) 1-LB Bag Granular;(1) 2 LB Acid Neutralizer;(1) Pair of Goggles;(1) Pair of Neoprene Gloves;(1) Respirator;(1) Scoop and Scraper;(10) 15 in x 19 in Pads;(2) Disposal Bags
Dimensions (L x W) 14 x 18 in.