Learn More
Fisherbrand™ FSC SXT™ Oil Only Absorbent Pads
Catalog No. 19140663 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
31.5 gal.
Catalog No. Capacity (English)
19-140-663 31.5 gal.
1 options
Catalog No. 19-140-663
  • $169.00
    $136.65 / Case of 200
    Save $32.35
    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Oil absorbents are high performance, laminated, oil only sorbents designed to handle petroleum-based spills especially for high abrasion applications.
  • Oil absorbents are high performance, laminated, oil only sorbents designed to handle petroleum-based spills especially for high abrasion applications.
  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use.
  • Light weight - specifically designed for smaller leaks, drips and spills.
  • Oil absorbents are ideal for wiping down tools or component parts and they remain intact even when saturated with oil.
  • Perforations provide cost control and reduce waste by allowing you to use only what you need.
Specifications
Capacity (English) Sorbing 31.5 gal.
Color White
Capacity (English) 31.5 gal.
Material Polypropylene
Type Spill Control Pads
Includes 200 Pads/Cs.
Dimensions (L x W) 19 x 15 in.