Learn More
Fisherbrand™ FSC Xtra Tough Universal Absorbent Pads
Catalog No. 19140660 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
31.5 gal.
Catalog No. Capacity (English)
19-140-660 31.5 gal.
1 options
Catalog No. 19-140-660
  • $181.00
    $158.65 / Case of 200
    Save $22.35
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Tough absorbents feature laminated coverstock - ideal for applications requiring a durable, non-linting and abrasion-resistant absorbent.
  • Tough absorbents feature laminated coverstock - ideal for applications requiring a durable, non-linting and abrasion-resistant absorbent.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids.
  • Light weight - specifically designed for smaller leaks, drips and spills.
  • Tough absorbents are made to handle virtually any liquid and have the extra strength required to allow for wringing and re-use.
  • Perforations provide cost control and reduce waste by allowing you to use only what you need.
Specifications
Capacity (English) Sorbing 31.5 gal.
Color Gray
Capacity (English) 31.5 gal.
Material Polypropylene
Type Spill Control Pads
Includes 200 Pads/Cs.
Dimensions (L x W) 19 x 15 in.