Learn More
Leica Biosystems FSC22 CLEAR 4FL.OZ 9 IN CASE
Catalog No. NC0242309
Encompass_Preferred
FSC 22 is a water soluble embedding compound used in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. It provides excellent sectioning consistency with minimal curling of sections at a working temperature of -20 °C. FSC 22 is available in clear or in light blue for better visualization of small specimens. It is highly recommended for surgical pathology laboratories.
Catalog No. NC0242309 Supplier Leica Biosystems Supplier No. 3801480
FSC 22 is a water soluble embedding compound used in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. It provides excellent sectioning consistency with minimal curling of sections at a working temperature of -20 °C. FSC 22 is available in clear or in light blue for better visualization of small specimens. It is highly recommended for surgical pathology laboratories.