Learn More
Aviva Systems Biology Corp FSCN2 Antibody - middle region (ARP59796_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59796_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FSCN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FSCN2. Alternate names: RFSN, RP30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-828