Learn More
Aviva Systems Biology Corp FSCN3 Antibody - middle region (ARP53741_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53741_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FSCN3. It was validated on Western Blot. FSCN3,fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotus purpuratus),FSCN3,fascin-3,fascin-3,testis fascinThe immunogen is a synthetic peptide directed towards the middle region of human FSCN3. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-878