Learn More
Aviva Systems Biology Corp FSCN3 Antibody - C-terminal region (ARP53740_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53740_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FSCN3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FSCN3. Alternate names: -ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-877