Learn More
New England Biolabs, Inc. FseI – 100 units
SDP
Catalog No. 50812298
Encompass
A restriction endonuclease that recognizes the sequence GG_CCGG^CC.
Catalog No. 50-812-298 Supplier New England Biolabs, Inc. Supplier No. R0588S
A restriction endonuclease that recognizes the sequence GG_CCGG^CC.