Learn More
New England Biolabs, Inc. FseI – 500 units
SDP
Catalog No. 50812297
Encompass
A restriction endonuclease that recognizes the sequence GG_CCGG^CC.
Catalog No. 50-812-297 Supplier New England Biolabs, Inc. Supplier No. R0588L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GG_CCGG^CC.