Learn More
Drg International Inc DRG FSH ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA1288
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of follicle stimulating hormone (FSH) in serum. Follicle-Stimulating Hormone (FSH) and Luteinizing Hormone (LH) are intimately involved in the control of the growth and reproductive activities of the gonadal tissues, which synthesize and secrete male and female sex hormones through a negative feedback relationship. The DRG FSH ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on a FSH molecule.
Catalog No. 50-187-1247