Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2492MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: FSHR/1400. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Uterine Carcinoma. Cellular Staining: Cytoplasmic ;Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length FSHR protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 75kDa.Chromosome: 2p21-p16. Gene Symbol: FSHR. Gene ID: 2492. Ensemble: ENSG00000170820. Uniprot: XP_011531035.
Catalog No. 50-188-4713