Learn More
Medchemexpress LLC FSL-1 | 322455-70-9 | 99.22% | 1666.16 | 5 MG
SDP
Catalog No. 5000422906
Encompass_Preferred
FSL-1 is a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist that enhances resistance to experimental HSV-2 infection. It also reduces HSV-2 replication in human vaginal epithelial cells and induces significant resistance to experimental genital HSV-2 infection through a specific cytokine response profile. Additionally, FSL-1 enhances phagocytosis of bacteria by macrophages via a Toll-like receptor 2-mediated signaling pathway.
  • Enhances resistance to experimental HSV-2 infection.
  • Reduces HSV-2 replication in human vaginal epithelial cells.
  • Induces significant resistance to experimental genital HSV-2 infection.
  • Enhances phagocytosis of bacteria by macrophages.
Catalog No. 50-004-22906 Supplier Medchemexpress LLC Supplier No. HYP20365MG
FSL-1 is a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist that enhances resistance to experimental HSV-2 infection. It also reduces HSV-2 replication in human vaginal epithelial cells and induces significant resistance to experimental genital HSV-2 infection through a specific cytokine response profile. Additionally, FSL-1 enhances phagocytosis of bacteria by macrophages via a Toll-like receptor 2-mediated signaling pathway.
  • Enhances resistance to experimental HSV-2 infection.
  • Reduces HSV-2 replication in human vaginal epithelial cells.
  • Induces significant resistance to experimental genital HSV-2 infection.
  • Enhances phagocytosis of bacteria by macrophages.