Learn More
AdipoGen FSL-1
SDP
Supplier:  AdipoGen AGCP30009C100
Encompass_Preferred
Chemical. CAS 322455-70-9. Formula C84H140N14O18S. MW 1666.2. Synthetic. FSL-1 Pam2CGDPKHPKSF is a synthetic lipoprotein that represents the N-terminal part of the 44kDa lipoprotein LP44 of Mycoplasma salivarium. Stimulator of TLR2/TLR6. Inducer of TNF-alpha production in macrophages. Upregulates proinflammatory cytokines. Activator of the proinflammatory transcription factor NF-kappaB.
Catalog No. 50-256-2455