Learn More
Medchemexpress LLC HY-P2036A 100ug Medchemexpress, FSL-1 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP2036A100UG
Encompass_Preferred
Medchemexpress, HY-P2036A 100ug FSL-1 (TFA) CAS: FSL-1 TFA, a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist, enhances resistance to experimental HSV-2 infection. FSL-1 TFA induces MMP-9 production through TLR2 and NF-κB/AP-1 signaling pathways in monocytic THP-1 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3079