Learn More
Medchemexpress LLC FSL-1 TFA | 99.8% | 1666.16 | 5 MG
SDP
Catalog No. 5000423541
Encompass_Preferred
FSL-1 TFA is a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist that enhances resistance to experimental HSV-2 infection. It induces MMP-9 production through TLR2 and NF-κB/AP-1 signaling pathways in monocytic THP-1 cells.
  • Targets toll-like receptor 2/6.
  • Enhances resistance to experimental HSV-2 infection and significantly reduces HSV-2 replication in human vaginal epithelial cells.
  • Induces MMP-9 production in monocytic THP-1 cells.
  • Activates the MAP kinase/NF-κB signaling pathway.
  • Protects against genital HSV-2 challenge in mice.
Catalog No. 50-004-23541 Supplier Medchemexpress LLC Supplier No. HYP2036A5MG
FSL-1 TFA is a bacterial-derived toll-like receptor 2/6 (TLR2/6) agonist that enhances resistance to experimental HSV-2 infection. It induces MMP-9 production through TLR2 and NF-κB/AP-1 signaling pathways in monocytic THP-1 cells.
  • Targets toll-like receptor 2/6.
  • Enhances resistance to experimental HSV-2 infection and significantly reduces HSV-2 replication in human vaginal epithelial cells.
  • Induces MMP-9 production in monocytic THP-1 cells.
  • Activates the MAP kinase/NF-κB signaling pathway.
  • Protects against genital HSV-2 challenge in mice.