Learn More
Medchemexpress LLC Fsllry-nh2 tfa | 245329-02-6 | 99.7% | 910.98 | 10 MG
SDP
Catalog No. 5000423858
Encompass_Preferred
FSLLRY-NH2 TFA is a protease-activated receptor 2 (PAR2) inhibitor used for research purposes. Treatment with FSLLRY-NH2 has been shown to significantly improve neurological outcome and reduce the number of degenerating hippocampal neurons after ACA (asphyxial CA).
  • Protease-activated receptor 2 (PAR2) inhibitor
  • Significantly improves neurological outcome after cardiac arrest in rats
  • Reduces degenerating hippocampal neurons after asphyxial CA
  • For research use only
Catalog No. 50-004-23858 Supplier Medchemexpress LLC Supplier No. HYP1260A10MG
FSLLRY-NH2 TFA is a protease-activated receptor 2 (PAR2) inhibitor used for research purposes. Treatment with FSLLRY-NH2 has been shown to significantly improve neurological outcome and reduce the number of degenerating hippocampal neurons after ACA (asphyxial CA).
  • Protease-activated receptor 2 (PAR2) inhibitor
  • Significantly improves neurological outcome after cardiac arrest in rats
  • Reduces degenerating hippocampal neurons after asphyxial CA
  • For research use only