Learn More
Apexbio Technology LLC FSLLRY-NH2 TFA 245329-02-6 (free base) 25mg
SDP
Catalog No. 5000088528
Encompass_Preferred
FSLLRY-NH2 TFA is a synthetic peptide antagonist targeting protease-activated receptor 2 (PAR-2) It is designed to block PAR-2 activation thereby inhibiting intracellular signaling cascades such as calcium mobilization and MAP kinase activation that underlie inflammatory responses nociceptive transmission and immune modulation FSLLRY-NH2 TFA exerts its biological activity primarily through competitive binding to PAR-2 preventing receptor activation and downstream signaling events Based on these pharmacological properties FSLLRY-NH2 TFA holds research potential in the study of inflammation-associated disorders and pain pathways
Catalog No. 50-000-88528 Supplier Apexbio Technology LLC Supplier No. B570325
FSLLRY-NH2 TFA is a synthetic peptide antagonist targeting protease-activated receptor 2 (PAR-2) It is designed to block PAR-2 activation thereby inhibiting intracellular signaling cascades such as calcium mobilization and MAP kinase activation that underlie inflammatory responses nociceptive transmission and immune modulation FSLLRY-NH2 TFA exerts its biological activity primarily through competitive binding to PAR-2 preventing receptor activation and downstream signaling events Based on these pharmacological properties FSLLRY-NH2 TFA holds research potential in the study of inflammation-associated disorders and pain pathways